In some cases, you might need to use more than one processing method in the sequence. For example, the sequest.pmd method runs the Daemon utility with a parameter file containing a simple Sequest HT workflow, and the export.pmd method runs the utility with an export workflow.
Procedure
- In the Sequence Setup view, select File > New.
- The New Sequence Template dialog box opens.
- Enter the appropriate values in each of the boxes.
- In the Bracket Type area, select the None option.
- You can now change the processing methods individually for each sample.
New Sequence Template with the selection of None for the bracket type Sequence with two different processing methods - Select OK.
- In this example, the Xcalibur data system starts two different workflows (performing a Sequest HT search and exporting a raw data file) for the recorded raw data files in the Proteome Discoverer application.
Two workflows in the job queue started by two different processing methods