You can use the Job Queue page in the Daemon utility window to monitor the execution of the jobs that you submit. It performs the same function as the job queue on the Proteome Discoverer interface. For information about the features of the job queue on the Proteome Discoverer interface, refer to the Job queue Help topic.
A progress bar displays the progress of the overall batch processing. This progress bar is visible only if you have started batch jobs.
Procedure
- Select the Job Queue tab of the Daemon utility window.
Job Queue page of the Daemon utility for batch processing