The Daemon utility can connect to a remote computer running the Magellan server so that you can perform searches on multiple raw data files from multiple samples or a single sample on a remote computer. It can also connect to a local server.
Procedure
- Select the Configuration tab in the Daemon utility window.
- From the Host list, select the name of the server that you want to use, or enter the server name.
- You must connect the utility to a computer running the Magellan server. Your local host is the default server, that is, the computer that you are working on. To connect remotely to the Magellan server, see Log in to a remote server.
- In the User box, enter the user’s login name on the server.
- In the Access Token box, enter or paste in your Access Token. You can copy the Access Token from the Daemon Access Token dialog box, see Create Daemon user and token.
Configuration page of the Proteome Discoverer Daemon utility - Select Apply to activate the new settings.
- To return to the previous settings, select Reset.