You can start the Proteome Discoverer Daemon utility on the command line or in a window. To run it on the command line, see Run the Daemon utility from the command line.
Procedure
  1. Start the utility in Windows by choosing Start > All Programs > Thermo Proteome Discoverer release_number > Proteome Discoverer Daemon release_number or by selecting Daemon on your desktop.
  2. After the utility window opens, connect to a computer that is running the Proteome Discoverer application.