Create Daemon user and token - Create Daemon user and token - Proteome Discoverer 3.1 User Guide - Proteome Discoverer Software - Create Daemon user and token - Proteome Discoverer Software - Proteome Discoverer Software - topic
Proteome Discoverer 3.1 User Guide
- Product Tree
- Desktop Software > Proteome Discoverer Software
- Ardia Platform > Software > Proteome Discoverer Software
- Document Type
- User Guide
- Software version
- 3.1
- Open the Proteome Discoverer application on the remote server.
- From the application tool bar, select Administration > Configuration.
- In the Configuration area, select Server Settings > Discoverer Daemon.
- Configuration page for Discoverer Daemon
- Select Create Daemon User and Access Token.
- The Daemon Access Token dialog box opens.
- Enter a username in the New Daemon User field and select Add.
- The new username appears in the Daemon User dropdown and the Access Token populates.
- Daemon Access Token dialog box
- Select OK.