You can now create a workflow for the Daemon utility with just the processing workflow. Using both processing and consensus workflows is no longer required, however the option to use both is still available.
Procedure
- If you have not already done so, create a processing (and consensus (optional)) workflow on the remote Proteome Discoverer server.
- After you create the workflow, copy the workflow to the instrument computer where the Daemon utility is installed.