LCMS Features page columns
Column
Description
# Isotopes
Displays the number of isotopic peaks matched for this feature.
Apex RT [min]
Displays the retention time at the apex of the feature.
Area
Displays the area of the largest theoretical contained LCMS peak.
Charge
Displays the charge state of the precursor ion associated with the feature.
Checked
Indicates whether the item is selected.
File ID
Displays a short identifier of the input file in which the application detected the feature.
Intensity
Displays the peak height of the largest theoretical contained LCMS peak.
Left RT [min]
Displays the lower retention time boundary of the feature.
m/z [Da]
Displays the monoisotopic m/z of the feature.
Quan Info
Displays the status of the validity of this quantification result for the quantification. The possible values are:
(For precursor ion quantification)
  • No Quan Values: Indicates that the feature area is 0.
  • Rejected by Method: Indicates that the peptide was not used in the quantification.
  • Shared: Indicates that different PSMs are associated with a feature.
(For reporter ion quantification)
  • No Quan Values: Indicates that the feature area is 0.
  • Rejected: Indicates that the peptide was not used in the quantification.
  • Shared: Indicates that different PSMs are associated with a feature.
Quan Info Details
Displays the reason for rejection when the status in the Quan Info column is Rejected by Method. The possible reasons are:
  • Single Peak: Indicates that the feature consists of only a single
    LC/MS peak and that single peak features should not be used.
  • Missing Replicates: Indicates that not enough replicate files have detected features.
Right RT [min]
Displays the upper retention time boundary of the feature.
Score
Displays the feature score for the feature.
Score Deviations
Displays the score deviation for the feature.
Workflow ID
Displays the identification of the workflow that the application detected the feature in.