The following table lists the parameters in the Specialized Traces view.
Command or Option | Description |
|---|---|
Checked | Indicates whether the item is selected. |
Description | Displays a description of the trace, the settings, or both. |
File ID | Displays the short identifier of the input file that the trace belongs to. |
Spectrum File | Displays the name of the input file that the application retrieved the trace from. |
Trace/Detector Type | Displays the trace type or analog detector type. |