The following views are available on the Specialized Traces page.
- Chromatogram Traces view (see Using the Chromatogram Traces view).
The Chromatogram Traces view used on this page can verify that the Minora algorithm and the retention-time alignment worked correctly. For example, the following figure compares two TIC traces from different files, which are highlighted in blue on the Specialized Traces page. Their chromatographic peaks, shown in turquoise and gold colors, overlay each other closely in the Chromatogram Traces view, indicating that the processing performed well.