PSMs are peptide sequences assigned to MS/MS spectra by a search engine. A PSM is different from a peptide group in that many PSMs might be linked to the same peptide group.
PSMs are peptide sequences assigned to MS/MS spectra by a search engine. A PSM is different from a peptide group in that many PSMs might be linked to the same peptide group.