Some column headings include an indication of the search node. The indication includes the following:
- The letter represents the number of the MSF file used in the consensus workflow that generated the data. For example, the first MSF file used is A, the second MSF file used is B, and on.
- The number is the node number of the search engine node that was used in the processing workflow that was used to generate the MSF file. For example, if the search engine node is the third node added to the workflow, its number is 3; if it was the fourth to be added, its number is 4, and on.
Column headings can include the following:
- A search node name, for example, Sequest HT
- A search node name and a letter, for example, Sequest HT A
- A search node name, a letter, and a number, for example, Sequest HT A4
The setting of the Merge Mode parameter of the MSF Files node determines which items the application includes in the column headings.
The Analysis Settings page of the Result Summaries view displays the letters that represent the MSF files, for example, Processing Step A. This page also displays the node numbers in the workflow tree.
The following table describes the columns on the Peptide Groups page.
Column | Description |
|---|---|
# Missed Cleavages | Displays the number of cleavage sites in a peptide sequence that a cleavage reagent (enzyme) did not cleave. This number excludes cases where an amino acid, such as proline, inhibits the cleaving enzyme. For example, if proline resides next to lysine or arginine, trypsin does not cleave the lysine or arginine. The application returns a 0 value when all peptides have been completely digested. |
# Protein Groups | Displays the number of protein groups containing the peptide. |
# Proteins | Displays the total number of identified proteins containing the peptide. |
# PSMs | Displays the total number of identified PSMs in the peptide group, including those redundantly identified. |
Abundance Ratios | Displays the abundance ratios as normal space values. This column appears when there are sample ratios defined in the analysis setup. |
Abundance Ratios (by Bio. Rep.) | Displays the abundance ratios of the biological replicates as normal space values. |
Abundance Ratios (log2) | Displays the abundance ratios as log2 ratios. |
Abundance Ratios (log2) (by Bio. Rep.) | Displays the abundance ratios of the biological replicates as log2 values. |
Abundances | Displays the abundances of the samples before scaling and normalization. |
Abundances (by Bio. Rep.) | Displays the abundance values of the biological replicates. |
Abundances (by Bio. Rep.) Counts | Displays the number of abundance values used to calculate the abundances of the biological replicates. |
Abundances (Grouped) | Displays the abundance values of the sample groups. A grouped abundance value is calculated as the arithmetic mean of all the replicate abundance values within a sample group. You can specify the sample grouping on the Grouping and Quantification page when you set up an analysis. This column appears when you group samples in the analysis setup and there is at least one sample group consisting of at least two samples. |
Abundances (Grouped) Counts | Displays the number of samples with detected abundance values used to calculate the abundance of the whole sample group. |
Abundances (Grouped) Standard Errors [%] | Displays the standard error of the abundance values of the samples in a sample group, normalized to the group’s median abundance. |
Abundances (Normalized) | Displays the normalized abundances values of the samples. This column appears when you set the Normalization Mode parameter of the Precursor Ions Quantifier node or the Reporter Ions Quantifier node to Total Peptide Amount or Specific Protein Amount. |
Abundances (Scaled) | Displays the normalized and scaled abundance values of the samples. This column appears when you set the Scaling Mode parameter of the Precursor Ions Quantifier node or the Reporter Ions Quantifier node to On All Average or On Controls Average. |
Abundances Counts | Displays the number of abundance values used to calculate the sample abundance. |
Annotated Sequence | Displays the identified peptide sequence with the flanking residues from the master protein. |
Charge (by Search Engine) | Displays the charge state of the peptide. |
Checked | Indicates whether the item is selected. |
Concatenated Rank (by Search Engine) | Displays the value for the “best” PSM found by the search engine. This value includes the rank of the decoy PSMs for the same spectra. |
Confidence | Indicates a confidence level associated with the peptide group at the top level:
|
Confidence (by Search Engine) | Indicates a confidence level associated with the peptide group at the top level:
|
Found in Files | Represents the best confidence of the PSMs of the protein that the application identified in the files:
This column appears only when you include the Data Distributions node in the consensus workflow and set its Show Found in Files parameter to True. |
Found in Fractions | Represents the best confidence of the PSMs of the protein that the application identified in the fractions. The colors of the boxes represent the following:
This column appears only when you include the Data Distributions node in the consensus workflow and set its Show Found in Fractions parameter to True. |
Found in Sample Groups | Represents the best confidence of the PSMs of the protein that the application identified in the sample groups:
This column appears only when you include the Data Distributions node in the consensus workflow and set its Show Found in Sample Groups parameter to True. |
Found in Samples | Represents the best confidence of the PSMs of the protein that the application identified in the samples:
This column appears only when you include the Data Distributions node in the consensus workflow and set its Show Found in Samples parameter to True. |
m/z [Da] (by Search Engine) | Displays the mass-to-charge ratio of the precursor ion, in daltons. |
Master Protein Accessions | Displays the unique identifiers (accessions) of all master proteins from all protein groups that include the given peptide sequence. This column appears when you use the Protein Grouping node in the consensus workflow. |
Master Protein Descriptions | Displays a description of the master proteins. |
Modifications | Displays all the PTMs identified for the given peptide. The sites are displayed only for fixed modifications or for variable modifications when the confidence level is above the threshold set by the IMP-ptmRS node. They are indexed to the local sequence of the peptide, not to the master protein. |
Modifications (All Possible Sites) | Displays all modifications found in the PSMs of a peptide group. They are indexed to the local sequence of the peptide, not to the master protein. |
Modifications in Master Proteins | Displays the positions of the modifications in the master proteins only if the confidence level is above the threshold set by the IMP-ptmRS node. |
Channel Occupancy [%} | Displays the percentage of the quan channels that have non-zero abundance for the peptide. This value is calculated by the Reporter Ions Quantifier node. |
Modifications in Master Proteins (All Sites) | Displays the positions of the modifications in the master proteins with confidence levels returned by the IMP-ptmRS node. |
Percolator PEP (by Search Engine) | Displays the posterior error probability (PEP) of the best PSM of the peptide group that the search node identified. This column appears when the consensus workflow includes the Percolator node. For information on how this node calculates the posterior error probability, see Posterior Error Probability (PEP). |
Percolator q-Value (by Search Engine) | Displays the q-value of the best PSM of the peptide groups that the search node identified. This column appears when the consensus workflow includes the Percolator node. For information on how this node calculates the q-value, see q-value. |
Percolator SVMScore (by Search Engine) | Displays the maximum Percolator score for the identified peptide group. This column appears when the consensus workflow includes the Peptide Validator node. For information on this node, see Peptide Validator node. |
Positions in Master Proteins | Displays the positions of the peptide in the master protein. |
Protein Accessions | Displays the unique identifiers (accessions) of all proteins from all protein groups that include the given peptide sequence. The identifiers displayed in the Protein Group Accessions column are the same as those displayed in the Accession column on the Proteins and PSMs pages. |
PSM Ambiguity | Displays the grouping status of the PSM in the peptide group. This column can display the following categories:
For more information on the protein group inference process that the application uses to assign the grouping status of PSMs, see Understanding protein grouping. |
Quan Info | Indicates why the application used or did not use a peptide in quantification. The possible reasons are:
(For reporter ion quantification)
(For precursor ion quantification)
|
Quality PEP | Displays the posterior error probability (PEP) for the identified peptide group. This column appears when the consensus workflow includes the Peptide Validator node. For information on this node, see Peptide Validator node. |
Quality q-value | Displays the q-value for the identified peptide group. This column appears when the consensus workflow includes the Peptide Validator node. For information on this node, see Peptide Validator node. |
Rank (by Search Engine) | Displays the ordering of peptides by rank. |
RT [min] (by Search Engine) | Displays the retention time when the peptide was observed, in minutes. |
Search Engine Rank (by Search Engine) | Displays the original rank assigned by the search engine for all PSMs and peptide groups. |
Sequence | Displays the sequence of amino acids that compose the peptide in the peptide group. |
Sequence Length | Displays the number of amino acids in a sequence. |
SVM_Score | Displays the maximum Percolator score for the identified peptide group. |
Theo. MH+ [Da] | Displays the mass calculated from the sequence, the amino acid masses, and the defined modification masses, as opposed to the measured mass. |
DCn (by Search Engine) | Displays the normalized score difference between the currently selected PSM and the highest-scoring PSM for that spectrum. |
DM [ppm] (by Search Engine) | Displays the difference between the theoretical mass of the peptide and the experimental mass of the precursor ion, in parts per million. |
Dm/z [Da] (by Search Engine) | Displays the difference between the measured charged mass (m/z [Da]) and the theoretical mass of the same charge (z). |
DScore (by Search Engine) | Displays a measure of the difference between the top two scores for the peptides identified by that spectrum. The application calculates this score as follows: DScore = Score(Rank N Peptide) – Score(Rank 1 Peptide)/Score(Rank 1 Peptide). For more information about the delta score calculation, see The Delta Score column. |
Contaminant | Displays an X symbol next to the proteins marked as contaminants in the searched FASTA file or files. This column appears when the consensus workflow includes the Protein Marker node. For more information, see Protein Marker node. |
Ratio columns | Display the corrected ratio of the intensity of the reporter ion in a sample to the intensity of the reporter ion in the control sample. |
XCorr search_engine_number node_number | Scores the number of fragment ions that are common to two different peptides with the same precursor mass and calculates the cross-correlation score for all candidate peptides queried from the database (Sequest HT searches only). |
Quan Usage | Shows which peptide abundances are used for protein quantification.
|
Species Map | Extracts from the FASTA database the species names for proteins and displays and annotates them as colored entries in a distribution map. This column appears only when you include the Protein Marker node in the consensus workflow and set its As Species Map parameter to True. |
Species | Extracts from the FASTA database the species names for proteins and displays and annotates them as semicolon-separated text. This column appears only when you include the Protein Marker node in the consensus workflow and set its As Species Names parameter to True. |
P-Value | Displays the p value for the sample group calculated by running the Tukey HSD test (post hoc) after an analysis of variance (ANOVA) test. For more information on p-values, see Calculate p-values and adjusted p-values for quantification results. The p value is a number between 0 and 1. If the null hypothesis states that there is no difference between the sample groups for the variance tests, one of the following pertains:
The color-coding of the p-values is as follows: |
Adj. P-Values | Displays the p-values adjusted by using the Benjamini-Hochberg correction for the false discovery rate. For more information on p-values, see Calculate p-values and adjusted p-values for quantification results. The color-coding of the adjusted p-values is as follows: |