The following table describes the commands and filters available in the Display Filter pane.
Parameter | Description |
|---|---|
Left pane | Lists the main tables (pages) included in the current result file. |
ON/OFF button | Use to turn on or turn off the conditions for the associated main table. |
Indicator icons | Gray—Indicates that the table is unfiltered. Green—Indicates that a filter has been applied to the table. Yellow—Indicates that the table filter is off. |
Right pane | Displays the filter settings for the selected table. You can modify these. |
AND or OR | Specifies the logical connection between properties or groups. |
Add Group | Adds a group. |
Add Property | Adds a property. |
Buttons | |
Loads the selected filter set. | |
Opens the Export Filter dialog box so that you can save the filter set as a file. | |
Removes the selected filter from the Display Filter pane. | |
Removes all filters and conditions from the Display Filter pane. | |
Apply | Applies the filter settings and conditions to the data in the results report. |