You can save a filter set as the default filters of a result file. To do this, you set the display filters, save them to a .filterSet file, and then apply them to a newly generated result file by using the Display Settings node in the consensus workflow.
If you load a filter into a report that does not contain quantification information and therefore has no Heavy/Light column, this condition is unavailable, so it appears with a line drawn through it.
Loading a quantification filter into a report containing no quantification data
The application still preserves such conditions in the internal representation of the filter but ignores them for the current report.