You can use the application’s display filters to refine the results in the result file.
Procedure
- Open a result file, and select on the tab of the page that you want to view.
- Select View > Display Filter, or select the Display Filter icon.
- The dockable Display Filter pane opens at the top of the results file.
- Set the first condition as follows:
- Select Add Property, and then select a property from the menu.
- Select the operator (red) box, and then select an operator for the condition from the menu.
- For numerical values, the default operator is Is Equal To. For textual values, the default operator is Contains.
- In the values box, enter a value.
- (Optional) To add another filter, select Add Property.
- Another level of filtering appears above the original level, and a new Add Property box appears under the original.
NOTE
Multiple conditions are arranged as siblings of an And box, which means that both conditions must apply to display a particular item. Change this box to Or if only one of the conditions applies.