A single or multiconsensus result file displays a list of PSMs and proteins identified by the search engine that is specified in the processing workflow. You can apply the application’s extensive set of editable display filters to any of the pages (tables) in the result file to refine the results in the file.
Filtering your search results reduces the complexity of the display, allowing for a quicker and more productive analysis.
You can save your filters as a filter set (.filterset file) for reuse.