The Amino Acids page shows the amino acids that form a PSM. It is contained in the result file but is not immediately visible when you open the file. It displays the amino acids that the modification of a selected PSM can modify.
Procedure
- In the result file, select the PSMs tab.
- On the PSMs page, select a PSM, and then select Show Associated Tables.
- On the page associated with the PSM, select the Found Modifications tab if it is not already selected.
- On the Found Modifications page, select the modification and then select Show Associated Tables.
- On the page associated with the modification, select the Amino Acids tab.