The Decoy PSMs page opens in the result file when the processing workflow includes the Percolator node or the Target Decoy PSM Validator node. The Decoy PSMs page displays PSMs that pass the FDR threshold set in the Target Decoy PSM Validator or Percolator nodes.
NOTE
This table is hidden by default.