The Decoy Peptide Groups page displays the peptide sequences that pass the FDR threshold set in the Target Decoy PSM Validator node or the Percolator node.
To add this page to the result file, include the Percolator node or the Target Decoy PSM Validator node in the processing workflow.
NOTE
This table is hidden by default.
Decoy Peptide Groups page