The Decoy Protein Groups page opens in the result file when the processing workflow includes the Percolator node or the Target Decoy PSM Validator node. The Decoy Protein Groups page displays the protein groups from the decoy database that pass the FDR threshold set in the Target Decoy PSM Validator node or the Percolator node.
NOTE
This table is hidden by default.
Decoy Protein Groups page