The Mass Recalibrations page of the result file shows how well the recalibration performed for all the input files. If the value in the RMSE column is low, the recalibration performed well; if it is high, the mass recalibration performed poorly. The Mass Recalibrations page can only be seen as an associated table of the Input Files page.