The result file includes the Result Statistics page when the consensus workflow includes the Result Statistics node. This page displays statistics for all columns containing numerical values on the Protein Groups, Proteins, Peptide Groups, PSMs, and MS/MS Spectrum Info pages. The Result Statistics page does not display statistics for the columns on the Annotated Modifications page.
You can use the Result Statistics page to quickly assess the quality of your experiment. For example, a properly performed tryptic digestion displays only peptides with no missed cleavages.
For information on the Result Statistics node, see the Result Statistics node in the Proteome Discoverer Help.
Result Statistics page