The LCMS Features page lists the LCMS features detected in all the raw data files. You can use this page to see the LCMS features only associated with a consensus feature.
The LCMS Features page is available only when you perform label-free quantification or precursor ion quantification.
NOTE
The table is hidden by default.
LCMS Features page