The Consensus Features page lists the consensus features for the analysis and shows the calculated abundances for those features across all the data files.
The Consensus Features page is available only when you perform label-free quantification or ion precursor quantification.