The Modification Sites page lists all PTM sites found in the result. Each modification in the result table includes modification type, site location probability, position on peptide and protein sequence, and the local sequence motif.
The Modification Sites node generates the Modification Sites page in the results.