The term PSM stands for a peptide spectrum match. The PSMs page lists all the PSMs found in a sample. Use the PSMs page to perform most of your tabular and graphical analyses. You can also access various detailed views for peptide matches.
The term PSM stands for a peptide spectrum match. The PSMs page lists all the PSMs found in a sample. Use the PSMs page to perform most of your tabular and graphical analyses. You can also access various detailed views for peptide matches.