The Peptide Groups page displays all the grouped peptides in the search results. This page opens only if you used the PSM Grouper node in the consensus workflow.
By default, the peptides on the Peptide Groups page are sorted by confidence and q-value, if those values are available. If they are not available, the items are sorted by confidence and the score of the search node used.
Peptide Groups page