The Proteins page lists all the proteins identified during a search. You can examine the search results in terms of protein identification and access more details about the peptide identifications and corresponding information from the search input. The Proteins page provides detailed tabular information, a shortcut menu, and access to the peptide information. The page is displayed by default when you first open a result file.