You can remove search results from a study.
NOTE
Removing search results from a study does not remove files from the hard drive.
Procedure
  1. In an open study, select the Analysis Results tab if it is not already selected.
  2. Select the row of results that you want to delete
  3. Do one of the following:
  4. Select Remove Files, and then select Yes in the dialog box that opens.
  5. The selected results are deleted.
  6. Select Open Containing Folder, and then delete the results in the results file that opens.