The following table describes the parameters in the Create New Quantification Method dialog box.
Parameter | Description |
|---|---|
From Factory Defaults | Uses the settings from one of the default methods that come with a new application installation. |
From Existing Method | Uses the settings from the existing quantification methods shown in the Quantification Methods view. |
From Scratch | Uses the Precursor Ion Quan Method template so that you can build a new precursor ion quan method from scratch. |
Create | Opens the Quantification Method Editor dialog box using the method that you specified. |