The following table lists the parameters that you can use to specify the location and number of the FASTA indexes stored.
Parameter | Definition |
|---|---|
FASTA Index Directory | (Read-only) Specifies the path and name of the current directory where the FASTA indexes are stored. Default location: C:\ProgramData\Thermo\Proteome Discoverer 3.1\FastaDatabases |
New Directory | Opens the Browse for Folder dialog box so that you can specify the new path and name of the directory to store the FASTA indexes in. |
Maximum Number of FASTA Indexes | (Read-only) Specifies the current maximum number of FASTA indexes allowed. Default: 30 |
New Maximum Number of FASTA Indexes | Specifies the maximum number of FASTA indexes allowed. Default: 30, maximum: 50 |