The following table lists the parameters that you can use to configure the remote server that the Proteome Discoverer Daemon utility connects to.
Parameter | Definition |
|---|---|
Current File Directory | Specifies the location where you want to store the output files. Default location: C:\ProgramData\Thermo\Proteome Discoverer 3.1\PublicFiles |
New Directory | Opens the Browse for Folder dialog box so that you can locate the user-named folder that is in the PublicFiles folder on the server where you want to store the output files. |