The following table lists the parameters that you can use to specify the number of processing workflows and consensus workflows to execute at the same time.
Parameter | Definition |
|---|---|
Max. Number of Processing Workflows in Parallel Execution | Specifies the number of processing workflows to execute in parallel. Default: 1; range: 1–half the number of dual-processor cores on your computer For example, if your computer has a dual-core processor with hyperthreading, it has four logical cores. In this case, the value of the Max. Number of Processing Workflows in Parallel Execution parameter is 2 (half of 4). |
Max. Number of Consensus Workflows in Parallel Execution | Specifies the number of consensus workflows to execute in parallel. Default: 1; range: 1–half the number of dual-processor cores on your computer For example, if your computer has a dual-core processor with hyperthreading, it has four logical cores. In this case, the value of the Max. Number of Consensus Workflows in Parallel Execution parameter is 2 (half of 4). |