The following table lists the parameters for storing spectrum library files.
Parameter | Definition |
|---|---|
Temporary Directory | (Read-only) Displays the location of the current directory where spectrum libraries are stored. TIP: Use a local drive with at least 100 Gb of free space. Default location: C:\ProgramData\Thermo\Proteome Discoverer 3.1\SpectrumLibraries |
New Directory | Specifies the location in which to store the spectrum library files. |