The following table lists the parameter that you can use to configure the Minora Feature Detector node.
Parameter | Definition |
|---|---|
# Parallel Tasks | Specifies the number of feature creation tasks that the Minora Feature Detector node can start in parallel. If you set this parameter to 0, the node starts as many tasks as possible. Range: 90–no maximum; default: 0 |