The following table lists the parameters that you can use to configure the IMP-ptmRS node.
Parameter | Definition |
|---|---|
Random Seed | Specifies the seed value that defines the starting point for the random number generator in the IMP-ptmRS node. To limit the influence of the isoform order during peak depth optimization, you can randomly shuffle the order.
Range: –1–no maximum; default: –2 |
Maximum Search Engine Rank | Specifies the highest search engine rank of a peptide to be considered for site localization. Range: 1–no maximum; default: 5 |
Minimum Main Score | Specifies the minimal main score of a peptide to be considered for site localization. Range: 9–no maximum; default: 0 |
Maximum Number of Threads | Specifies the maximum number of threads used for scoring. Range: 0–no maximum; default: 0 |
Modification Configuration | Determines whether the IMP-ptmRS node performs additional modification configurations on items such as diagnostic ions, neutral losses, targets, or fragment ion types. |