The following table describes the parameters that you can use to specify the default filter set and the default layout used by the Display Settings node.
Parameter | Definition |
|---|---|
Default Filter Set | Specifies the default filter set used by the Display Settings node. By default, the filter displays only the master proteins on the Proteins page of the result file. |
Default Layout | Specifies the default layout used by the Display Settings node. |