The following table lists the parameters for configuring access to the Annotation Server to retrieve protein annotations.
Parameter | Definition |
|---|---|
Annotation service URL | Specifies the path and name of the annotation service, which is the following: |
Number of Attempts to Submit the Annotation Request | Specifies the number of times that the application tries to submit the search when the server is busy. Default: 3 |
Time Interval Between Attempts to Submit the Annotation Request [sec] | Specifies the interval of time, in seconds, that elapses between attempts to submit a search when the Annotation Server is busy. Default: 90 |
Timeout of the Annotation Request [min] | Specifies the amount of time, in minutes, that elapses before the application ceases to try to access the Annotation Server when the server is busy. Default: 15 |