These settings are related to studies and analyses. The settings are active by default.
Procedure
- Select Tools > Options.
- In the Options dialog box, select Study Options.
- (Optional) Do any of the following:
- Select or clear the Auto Save When Starting Analysis option.
- If you select this option, the application automatically saves the contents of a study when you start analysis.
- Select or clear the Enable Study Variables Without Multiple Values option.
- If you select this option, you may select all variables (not just those with multiple values) on the Grouping & Quantification page.
- Select or clear the Show Warning When Running a Quan Analysis Without Specified Ratios option.
- If you select this option, the application displays a warning if you start an analysis without creating any ratios on the Grouping & Quantification page.
- Select OK, or select the close button x in the upper right corner to close the Options dialog box.