The following table describes the parameters available in the Annotation Aspects view on the Administration page.
Parameter | Description |
|---|---|
Implements any changes that you make to the ID, Read Only, or Active columns. | |
Opens the Annotation Aspects Editor so that you can define a new annotation aspect. | |
Removes an annotation aspect from the table on the Annotation Aspects view. | |
Opens the Annotation Aspects Editor so that you can change the settings of an existing annotation aspect. | |
Imports an XML file containing annotation aspects and displays it in the table on the Annotation Aspects view. | |
Places the selected annotation aspects in an XML file. | |
Updates an annotation database before you define a new annotation aspect for the first time after the installation of the application. | |
ID | Displays the sequential identification number of the annotation aspect. |
Read Only | Indicates whether an annotation aspect can be changed or deleted. (For example, the Edit and Delete buttons in the toolbar above the list are read-only.) User-defined annotation aspects are not read-only. |
Active | Determines whether the annotation aspect is displayed in the Protein Annotation node. |
Aspect Name | Displays the name of the annotation aspects. |
Position | Specifies the position of the colored rectangle that represents a group assigned to the annotation aspect. |
Color | Specifies the color that a group of an annotation aspect displays in the results report (result file). |
Description | Briefly describes the group of an annotation aspect. |