The following table describes the parameters available in the Annotation Aspect Editor.
Annotation Aspect Editor parameters
Parameter
Description
Aspect Name
Specifies the name of the annotation aspect.
Description
Briefly describes the annotation aspect.
Aspect definition area
Displays colored rectangles corresponding to the terms assigned to the annotation aspect.
Annotation Group
Contains terms that define a group.
Name
Specifies the name of the group.
New Group
Places a rectangle in the aspect definition area to represent the new group being defined.
Position
Specifies the position of a rectangle representing a group in the aspect definition area.
Color
Specifies the color that the group displays in the results report (.pdResult file).
Delete Group
Deletes a group.
Included Annotations
Lists the annotations assigned to a group.
Accession
Displays the accession number of the annotation.
Description
Briefly describes the annotation.
# Child Terms
Displays the number of child terms that the annotation has.
<
Assigns a term to an annotation group.
>
Removes a term from an annotation group.
Available Annotations
Lists the annotations in the selected database.
Annotation Database
Selects the database of annotations to display.
Accession
Displays the accession number of the annotation.
Description
Briefly describes the annotation.
# Child Terms
Displays the number of child terms that the annotation has.
Cancel
Discards any changes to the annotation aspect and closes the Annotation Aspect Editor.
Saves
Saves the annotation aspect.