The following table describes the parameters in the Cleavage Reagents view.
Parameter | Description |
|---|---|
Remove | Deletes the highlighted cleavage reagent. |
Apply | Saves any changes made to the cleavage reagents. |
Name | Specifies the name of the reagent used for the protein digestion. The options are as follows:
|
Cleavage Sites | Specifies the position (amino acid) at which to cleave the sequence. The options are as follows:
|
Cleavage Inhibitors | Specifies the amino acids that block cleavage when they are adjacent to the cleavage site. The options are as follows:
|
Offset | Specifies which side of the given amino acids the protease should cleave. The options are as follows:
|
Abbreviation | Specifies the user-defined abbreviation. The options are as follows:
|
Cleavage Specificities | Selects the cleavage specificity for enzymes. You can select more than one cleavage specificity. The available cleavage specificities are as follows:
|
Click here to add a new record | Opens a row in the reagents table so that you can enter the name and information about a new cleavage reagent. |