You can remove chemical modifications from the Chemical Modifications view.
Procedure
- Do one of the following:
- Select Administration > Maintain Chemical Modifications.
- Select Maintain Chemical Modifications
.
- The Chemical Modifications view opens.
- Select the row of the modification that you want to delete.
- Select Remove.
- In the Delete Row dialog box, select Yes.