You can create new chemical modifications and add them to the Chemical Modifications view. For example, you might have a new or experimental label that you want to add to the list of chemical modifications.
Procedure
- Do one of the following:
- Select Administration > Maintain Chemical Modifications.
- Select Maintain Chemical Modifications
.
- The Chemical Modifications view opens.
- At the top of the modification table, select Add.
- The Add Chemical Modification dialog box opens.
- At the top of the dialog box, do the following:
- In the Name box, enter the name of the modification.
- In the Abbreviation box, enter an abbreviation for the modifications.
- On the General page of the dialog box, enter the modification position, the delta mass, the chemical substitution, and the leaving group in the corresponding text boxes.
- In the Amino Acid Sites table, specify the target amino acids and the classification of the modification at that site as follows:
- To add a new row, select the lowest row in the amino acids sites table, and press Enter.
- To delete a row, select it and press Delete.
- If the chemical modification exhibits either a neutral loss or an immonium-type ion in the MS/MS spectrum, add this information on the Neutral Losses page and the Diagnostic Ions page, respectively.
- This feature is used by the Fragment Match Spectrum to label these diagnostic ions in the spectrum.
- To accept the new modifications, select OK.
- The Crosslinking tab is reserved for the definition of cross-linkers used by the XlinkX nodes for Proteome Discoverer.
NOTE
The XlinkX nodes must be purchased separately.