You can delete a spectrum library from the application.
NOTE
After you start the deletion of a file, you cannot cancel the deletion.
Procedure
- Select Administration > Maintain Spectrum Libraries.
- The Administration page opens with the Spectrum Libraries view.
- Select the row of the library that you want to delete.
- Select Remove.
- In the Remove Spectrum Libraries Databases dialog box, select OK.