If a spectrum library requires modification mapping, you must do that before exporting it. For more information, see Map unrecognized modifications.
Procedure
  1. Select Administration > Maintain Spectrum Libraries.
  2. Select a spectrum library, and then select Export.
  3. Select the folder that you want to export the library to.
  4. The Job Queue opens and shows the progress of the export.