You can download a spectrum library from a list of indexed, ready-to-use libraries by using the Download button. When you select one of these libraries, the application imports it automatically.
NOTE
You do not add spectrum libraries by using the Add button.
The Spectrum Library Download Server URL configuration parameter specifies the URL of the server that provides spectrum libraries for download.
Procedure
- Select Administration > Maintain Spectrum Libraries.
- Select Download.
- In the Spectrum Library Download dialog box, select the library that you want to import..
- Select Import.
- The Job Queue opens and shows the progress of the download.
- Wait until the Job Queue confirms that the download is complete, and then select Spectral Libraries.