To use a spectrum library, the application must register it. If you have a local spectrum library that you did not download directly into the application, you must import the library by using the Add button. During registration, the application:
- Transfers the file or archives it to the server
- Converts the library to an internal format, which includes the following:
- Extracting the library spectrum and peptide match annotations
- Annotating the library spectrum with the activation type and fragment match tolerance specified when the library was added
- Mapping modifications to known modifications
- Registers optional FASTA files contained in the library archive and generates mapping from the peptide matches in the library to the proteins in the FASTA file.
- Generates the target and decoy versions of the library in MSPepSearch binary format.
For information on downloading a ready-to-use spectrum library directly into the application, see Download a spectrum library.
Procedure
- Outside of the Proteome Discoverer application, download the appropriate spectrum libraries from the National Institute of Standards and Technology (NIST) at http://peptide.nist.gov or from Peptide Atlas at http://www.peptideatlas.org/speclib.
- In the application, select Administration > Maintain Spectrum Libraries.
- Select Add.
- In the Spectrum Library Import dialog box, do the following:
- For Library File, select the file selector icon, and select the spectrum library file.
- Select Open.
- The Spectrum Library Import dialog box opens.
- For Library Name, enter a name, or accept the default (that is, the entry for Library Name without the suffix).
- (Optional) Enter a number for Version.
- (Optional) Enter a description for Organism.
- Select the activation type from the Activation Type dropdown list..
- The activation type determines the fragment ion types used for annotating the fragment peaks in the library spectra.
- For Fragment Match Tolerance, enter a number and select the corresponding unit, Da or ppm.
- This value is used for annotating the fragment peaks in the library spectra.
- (Optional) For Collision Energy (Min-Max), enter a value for one or both fields.
- (Optional) Enter a comment.
Adding a Spectral Library - Select OK.
- The job queue in the Administration view displays the status each step of library creation. When the application finishes adding the spectrum library, it appears in the Spectrum Libraries view.
- If the application finds unknown modifications, you must manually map them before you can search the library or export it. See Map unrecognized modifications.