Use the Field Chooser to specify the columns that you want to display.
Procedure
- Select Field Chooser
.
- In the Field Chooser dialog box, select the checkboxes for the columns that you want to display in the Spectral Library view.
- The application updates the layout.