You can display a list of all the spectrum libraries that you registered in the Proteome Discoverer application.
Procedure
- Select Administration > Maintain Spectrum Libraries.
- The Spectrum Libraries view opens.
- The view displays the available spectrum libraries. The information, organized in columns, includes the file name, the file size, the number of proteins stored, and so on.
- For information on changing the columns in the display, see Select the columns to display.