The following table describes the parameters in the FASTA Indexes Options dialog box.
Parameters | Description |
|---|---|
FASTA Index Directory | Displays the path and name of the current directory where the FASTA indexes are stored. This parameter is read-only. Default: C:\ProgramData\Thermo\Proteome Discoverer 3.1\FastaDatabases |
New Directory | Opens the Browse for Folder dialog box so that you can specify the new path and name of the directory to store the FASTA indexes in. |
Maximum Number of FASTA Indexes | Specifies the current maximum number of FASTA indexes allowed. This parameter is read-only. |
New Maximum Number of FASTA Indexes | Specifies the new maximum number of FASTA indexes allowed. Default: 30 |
Reset | Resets the changes that you made in the FASTA Indexes Options dialog box to their values the last time the server started and prevents the server from implementing the changes when it starts next. |